Gene Transcripts

Gene Symbol
Nlk
Entrez Gene ID
497961
Gene Name
nemo like kinase
Gene Aliases
RGD1561602
Chromosome Location
Chr. 10: 65840052 - 65963932 on Build Rnor_6.0
UniGene ID
Rn.113514
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191924.1 NP_001178853.1 6, 7 2242
XM_006246945.2 XP_006247007.1 6, 7 2044

View Full Product Details