Gene Transcripts

Gene Symbol
Ttc30b
Entrez Gene ID
499814
Gene Name
tetratricopeptide repeat domain 30B
Gene Aliases
Chromosome Location
Chr. 3: 62758754 - 62761325 on Build Rnor_6.0
UniGene ID
Rn.87109
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127607.1 NP_001121079.1 1 1220
GenBank mRNA BC166739.1 - 1 1220

View Full Product Details