Gene Transcripts

Gene Symbol
Bpifb4
Entrez Gene ID
499925
Gene Name
BPI fold containing family B, member 4
Gene Aliases
Lplunc4, RY2G5
Chromosome Location
Chr. 3: 149424392 - 149449350 on Build Rnor_6.0
UniGene ID
Rn.117356
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109209.2 NP_001102679.2 8, 9 1148
GenBank mRNA X60660.1 - 6, 7 707

View Full Product Details