Gene Transcripts
- Gene Symbol
- Serf1
- Entrez Gene ID
- 502503
- Gene Name
- small EDRK-rich factor 1
- Gene Aliases
- RGD1565105
- Chromosome Location
- Chr. 2: 30345861 - 30352114 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_577983.5 |
XP_577983.3
|
2 |
353 |
|
View Full Product Details