Gene Transcripts

Gene Symbol
Ugt1a5
Entrez Gene ID
574523
Gene Name
UDP glucuronosyltransferase 1 family, polypeptide A5
Gene Aliases
UDPGT 1-5, Ugt1, Ugt1a6
Chromosome Location
Chr. 9: 95256628 - 95302822 on Build Rnor_6.0
UniGene ID
Rn.26489
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039549.1 NP_001034638.1 1 372
GenBank mRNA AF461734.1 - 1 381
AY435132.1 - 1 337

View Full Product Details