Gene Transcripts

Gene Symbol
LOC681193
Entrez Gene ID
681193
Gene Name
similar to DNA-directed RNA polymerase II 7.6 kDa polypeptide (RPB10) (RPB7.6) (RPABC5)
Gene Aliases
Chromosome Location
Chr. 4: 2512039 - 2513976 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006235860.2 XP_006235922.1 1 1255

View Full Product Details