Gene Transcripts

Gene Symbol
Lysmd4
Entrez Gene ID
681647
Gene Name
LysM, putative peptidoglycan-binding, domain containing 4
Gene Aliases
Chromosome Location
Chr. 1: 128199113 - 128204989 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003748880.3 XP_003748928.1 3 463
XM_008759576.1 XP_008757798.1 3 466

View Full Product Details