Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004292.2 | NP_004283.2 | 10 | 2373 | |
XM_011545399.2 | XP_011543701.1 | 10 | 2722 | ||
XM_011545400.2 | XP_011543702.1 | 10 | 2092 | ||
XM_011545401.2 | XP_011543703.1 | 10 | 2247 | ||
XM_017018587.1 | XP_016874076.1 | 10 | 2095 | ||
GenBank mRNA | AB209445.1 | - | 7 | 2276 | |
AK295365.1 | - | 9 | 1964 | ||
AK301430.1 | - | 7 | 1662 | ||
BC014417.1 | - | 10 | 2263 | 4549333 | |
BG163919.1 | - | 1 | 154 | ||
KU178481.1 | - | 10 | 2246 | ||
KU178482.1 | - | 10 | 2060 | ||
KU178483.1 | - | 4 | 513 | ||
L36463.1 | - | 10 | 2373 |