Gene Transcripts

Gene Symbol
Ube2ql1
Entrez Gene ID
679949
Gene Name
ubiquitin-conjugating enzyme E2Q family-like 1
Gene Aliases
Chromosome Location
Chr. 1: 36185916 - 36226276 on Build Rnor_6.0
UniGene ID
Rn.21006
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145163.1 NP_001138635.1 2 1371

View Full Product Details