Gene Transcripts

Gene Symbol
Plekhg1
Entrez Gene ID
679812
Gene Name
pleckstrin homology domain containing, family G (with RhoGef domain) member 1
Gene Aliases
Chromosome Location
Chr. 1: 40320531 - 40521395 on Build Rnor_6.0
UniGene ID
Rn.225479
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190999.1 NP_001177928.1 14 2762
XM_006227838.2 XP_006227900.1 15 6166
XM_006227841.2 XP_006227903.1 16 3097
XM_006227842.2 XP_006227904.1 16 3115
XM_006227844.2 XP_006227906.1 15 3891
XM_008758706.1 XP_008756928.1 15 3006
XM_008758707.1 XP_008756929.1 15 3009

View Full Product Details