Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_008770390.1 | XP_008768612.1 | 30 | 4504 | |
XM_008770391.1 | XP_008768613.1 | 30 | 4483 | ||
XM_008770392.1 | XP_008768614.1 | 29 | 4330 | ||
XM_008770393.1 | XP_008768615.1 | 25 | 5246 |