Gene Transcripts

Gene Symbol
Plp2
Entrez Gene ID
302562
Gene Name
proteolipid protein 2 (colonic epithelium-enriched)
Gene Aliases
A4-LSB
Chromosome Location
Chr. X: 15679254 - 15682653 on Build Rnor_6.0
UniGene ID
Rn.96974
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207601.1 NP_997484.1 3 371
GenBank mRNA BC061844.1 - 3 371
FQ216622.1 - 3 407
FQ220564.1 - 3 410

View Full Product Details