Gene Transcripts
- Gene Symbol
- NUP58
- Entrez Gene ID
- 9818
- Gene Name
- nucleoporin 58
- Gene Aliases
- NUP45, NUPL1, PRO2463
- Chromosome Location
- Chr. 13: 25301152 - 25349795 on Build GRCh38
- UniGene ID
- Hs.507537
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001008564.1 |
NP_001008564.1
|
6, 7 |
919 |
|
NM_014089.3 |
NP_054808.1
|
7, 8 |
955 |
|
XM_005266600.3 |
XP_005266657.1
|
7, 8 |
1299 |
|
XM_011535328.1 |
XP_011533630.1
|
7, 8 |
1299 |
|
XM_011535329.2 |
XP_011533631.1
|
8, 9 |
1385 |
|
XM_017020880.1 |
XP_016876369.1
|
9, 10 |
1467 |
|
GenBank mRNA |
AB007870.1 |
- |
7, 8 |
852 |
|
AK123955.1 |
- |
8, 9 |
789 |
|
AK302946.1 |
- |
6, 7 |
814 |
|
AK303409.1 |
- |
6, 7 |
723 |
|
BC001104.2 |
- |
7, 8 |
827 |
3504343 |
View Full Product Details