Gene Transcripts

Gene Symbol
Styx
Entrez Gene ID
100912536
Gene Name
serine/threonine/tyrosine interacting protein
Gene Aliases
Chromosome Location
Chr. 15: 19690091 - 19720683 on Build Rnor_6.0
UniGene ID
Rn.205855
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271541.1 NP_001258470.1 11 824

View Full Product Details