Gene Transcripts

Gene Symbol
Mcts2
Entrez Gene ID
689500
Gene Name
malignant T cell amplified sequence 2
Gene Aliases
Chromosome Location
Chr. 3: 148167190 - 148169029 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006235283.1 XP_006235345.1 1 1351

View Full Product Details