Gene Transcripts

Gene Symbol
Vom2r13
Entrez Gene ID
690593
Gene Name
vomeronasal 2 receptor, 13
Gene Aliases
Chromosome Location
Chr. 1: 64928503 - 64975956 on Build Rnor_6.0
UniGene ID
Rn.217646
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099489.1 NP_001092959.1 48

View Full Product Details