Gene Transcripts

Gene Symbol
PSMB10
Entrez Gene ID
5699
Gene Name
proteasome subunit beta 10
Gene Aliases
LMP10, MECL1, beta2i
Chromosome Location
Chr. 16: 67934504 - 67936877 on Build GRCh38
UniGene ID
Hs.9661
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002801.3 NP_002792.1 4 423
GenBank mRNA AK225658.1 - 4 396
AK298012.1 - 2 731
AK312208.1 - 4 383
BC017198.1 - 4 374 3143189
BC052369.1 - 4 361 4054165
BM845639.1 - 4 396
BP289065.1 - 4 423
BT019723.1 - 4 295
BT019724.1 - 4 295
CR456982.1 - 4 295
Y13640.1 - 4 383

View Full Product Details