Gene Transcripts

Gene Symbol
MXD3
Entrez Gene ID
83463
Gene Name
MAX dimerization protein 3
Gene Aliases
BHLHC13, MAD3, MYX
Chromosome Location
Chr. 5: 177305500 - 177312291 on Build GRCh38
UniGene ID
Hs.743332
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142935.1 NP_001136407.1 4, 5 774
NM_031300.3 NP_112590.1 4, 5 774
GenBank mRNA AF114834.1 - 4, 5 368
AK057034.1 - 4, 5 381
AK222579.1 - 4, 5 379
AK303397.1 - 4, 5 381
AK316468.1 - 4, 5 378
AL522759.3 - 4, 5 793
AL833959.1 - 1, 2 1420
BC000745.2 - 4, 5 363 2821596
BC032586.1 - 4, 5 378

View Full Product Details