Gene Transcripts

Gene Symbol
SLC22A31
Entrez Gene ID
146429
Gene Name
solute carrier family 22 member 31
Gene Aliases
Chromosome Location
Chr. 16: 89195756 - 89201674 on Build GRCh38
UniGene ID
Hs.447544
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242757.1 NP_001229686.1 8 1712
XM_011522897.2 XP_011521199.1 9 1760
XM_011522899.2 XP_011521201.1 9 1697
XM_017022962.1 XP_016878451.1 6 1362
XM_017022963.1 XP_016878452.1 6 3110
XM_017022964.1 XP_016878453.1 7 2266
XM_017022965.1 XP_016878454.1 6 3051
XM_017022966.1 XP_016878455.1 7 2889
GenBank mRNA AL137382.1 - 3 1478

View Full Product Details