Gene Transcripts
- Gene Symbol
- EPB41L5
- Entrez Gene ID
- 57669
- Gene Name
- erythrocyte membrane protein band 4.1 like 5
- Gene Aliases
- BE37, YMO1, YRT
- Chromosome Location
- Chr. 2: 120013028 - 120179121 on Build GRCh38
- UniGene ID
- Hs.369232
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001184937.1 |
NP_001171866.1
|
2, 3 |
365 |
|
NM_001184938.2 |
NP_001171867.1
|
2, 3 |
655 |
|
NM_001184939.1 |
NP_001171868.1
|
2, 3 |
365 |
|
NM_020909.3 |
NP_065960.2
|
2, 3 |
365 |
|
NR_135920.1 |
|
2, 3 |
365 |
|
XM_006712651.1 |
XP_006712714.1
|
2, 3 |
345 |
|
XM_006712652.1 |
XP_006712715.1
|
2, 3 |
660 |
|
XM_011511529.2 |
XP_011509831.1
|
2, 3 |
345 |
|
XM_017004567.1 |
XP_016860056.1
|
2, 3 |
201 |
|
GenBank mRNA |
AK023019.1 |
- |
2, 3 |
316 |
|
AK290895.1 |
- |
2, 3 |
275 |
|
BC032822.2 |
- |
2, 3 |
655 |
4827274 |
BC054508.1 |
- |
2, 3 |
300 |
5502663 |
View Full Product Details