Gene Transcripts

Gene Symbol
C5orf52
Entrez Gene ID
100190949
Gene Name
chromosome 5 open reading frame 52
Gene Aliases
Chromosome Location
Chr. 5: 157671237 - 157680176 on Build GRCh38
UniGene ID
Hs.720862
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017008909.1 XP_016864398.1 1 21

View Full Product Details