Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_032461.3 | NP_115850.2 | 2 | 393 | |
GenBank mRNA | AF098307.1 | - | 2 | 393 | |
AJ457796.1 | - | 2 | 393 | ||
BC034472.1 | - | 2 | 391 | 4824918 | |
BC137533.1 | - | 2 | 383 | 9021533 | |
BC137536.1 | - | 2 | 383 | 9021536 |