Gene Transcripts

Gene Symbol
OVOS2
Entrez Gene ID
144203
Gene Name
ovostatin 2
Gene Aliases
Chromosome Location
Chr. 12: 31114623 - 31206154 on Build GRCh38
UniGene ID
Hs.524331
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AL831947.1 - 3261
BC039117.1 - 8 1454
BC046190.1 - 6 1105
CR749395.1 - 5 3761

View Full Product Details