Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_011510199.1 | XP_011508501.1 | 2 | 1217 | |
XM_011510200.1 | XP_011508502.1 | 3 | 237 | ||
XM_011510201.1 | XP_011508503.1 | 4 | 423 | ||
XM_011510202.1 | XP_011508504.1 | 2 | 162 | ||
XM_017002956.1 | XP_016858445.1 | 3 | 1471 | ||
XM_017002957.1 | XP_016858446.1 | 3 | 1323 | ||
XM_017002958.1 | XP_016858447.1 | 3 | 1310 | ||
XM_017002959.1 | XP_016858448.1 | 4 | 383 | ||
XM_017002960.1 | XP_016858449.1 | 3 | 1471 | ||
XM_017002961.1 | XP_016858450.1 | 2 | 1217 | ||
XM_017002962.1 | XP_016858451.1 | 3 | 237 | ||
XM_017002964.1 | XP_016858453.1 | 2 | 1217 | ||
XM_017002965.1 | XP_016858454.1 | 1 | 546 | ||
GenBank mRNA | BX648271.1 | - | 1 | 2957 | |
BX649148.1 | - | 1 | 3759 | ||
CR627439.1 | - | 2 | 1168 | ||
CR627446.1 | - | 1 | 3269 |