Gene Transcripts

Gene Symbol
FAHD2B
Entrez Gene ID
151313
Gene Name
fumarylacetoacetate hydrolase domain containing 2B
Gene Aliases
Chromosome Location
Chr. 2: 97080738 - 97094882 on Build GRCh38
UniGene ID
Hs.567723
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320848.1 NP_001307777.1 6 916
NM_199336.2 NP_955368.1 5 790
XM_011510745.2 XP_011509047.1 6 906
XM_011510746.2 XP_011509048.1 6 1025
XM_011510747.2 XP_011509049.1 5 790
XM_011510748.2 XP_011509050.1 6 906
XM_017003470.1 XP_016858959.1 6 903
XM_017003471.1 XP_016858960.1 6 906
XM_017003472.1 XP_016858961.1 5 839
GenBank mRNA AL833869.1 - 4 590
BC064511.1 - 5 753 6644987

View Full Product Details