Gene Transcripts

Gene Symbol
Gsx2
Entrez Gene ID
14843
Gene Name
GS homeobox 2
Gene Aliases
Gsh-2, Gsh2
Chromosome Location
Chr. 5: 75075601 - 75077893 on Build GRCm38
UniGene ID
Mm.218752
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133256.2 NP_573555.1 1 247
GenBank mRNA BC119464.1 - 1 246
BC119465.1 - 1 246
BC134359.1 - 1 246
S79041.1 - 1 247

View Full Product Details