Gene Transcripts

Gene Symbol
Mcpt1
Entrez Gene ID
17224
Gene Name
mast cell protease 1
Gene Aliases
AV080368, Mcp-1
Chromosome Location
Chr. 14: 56017964 - 56020391 on Build GRCm38
UniGene ID
Mm.201549
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008570.1 NP_032596.1 4 434
GenBank mRNA BC100700.1 - 4 425
BC100701.1 - 4 425 40043850
BC100702.1 - 4 425 40043849
BC100703.1 - 4 425 40043848
S44609.1 - 4 427

View Full Product Details