Gene Transcripts

Gene Symbol
Mt4
Entrez Gene ID
17752
Gene Name
metallothionein 4
Gene Aliases
MT-IV
Chromosome Location
Chr. 8: 94137204 - 94139031 on Build GRCm38
UniGene ID
Mm.1536
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008631.2 NP_032657.1 3 263
GenBank mRNA AK131894.1 - 3 263

View Full Product Details