Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013647.2 | NP_038675.2 | 1, 2 | 156 | |
GenBank mRNA | AK002904.1 | - | 1 | 96 | |
AK010613.1 | - | 1 | 97 | ||
AK010641.1 | - | 1 | 97 | ||
AK011060.1 | - | 1 | 108 | ||
AK019400.2 | - | 1 | 95 | ||
AK083946.1 | - | 1 | 95 | ||
AK088413.1 | - | 1 | 96 | ||
AK088716.1 | - | 1 | 96 | ||
AK150615.1 | - | 1 | 89 | ||
AK150808.1 | - | 1 | 98 | ||
AK168207.1 | - | 1 | 95 | ||
AK169328.1 | - | 1 | 96 | ||
AV496563.1 | - | 1, 2 | 156 | ||
BC082286.1 | - | 1 | 75 | ||
BC090618.1 | - | 1 | 123 | 5713263 | |
BC115673.1 | - | 1 | 63 | 40104131 | |
BC115674.1 | - | 1 | 63 | 40104132 |