Gene Transcripts

Gene Symbol
Tmprss13
Entrez Gene ID
214531
Gene Name
transmembrane protease, serine 13
Gene Aliases
BC010843, Tpmrss13
Chromosome Location
Chr. 9: 45319071 - 45347581 on Build GRCm38
UniGene ID
Mm.394987
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013373.2 NP_001013391.2 11 1404
GenBank mRNA BC010843.1 - 3 253
BC042878.1 - 10 1140
BC085323.1 - 11 1389 30734093

View Full Product Details