Gene Transcripts

Gene Symbol
Rcor1
Entrez Gene ID
217864
Gene Name
REST corepressor 1
Gene Aliases
5730409O11, 6720480E22Rik, AU042633, D12Wsu95e, Rocr1, mKIAA0071
Chromosome Location
Chr. 12: 111038236 - 111115901 on Build GRCm38
UniGene ID
Mm.23808
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198023.2 NP_932140.1 9 874
GenBank mRNA AB093210.1 - 5 575
AK077445.1 - 3 315
BC042731.1 - 9 874 4936863

View Full Product Details