Gene Transcripts
- Gene Symbol
- CFDP1
- Entrez Gene ID
- 10428
- Gene Name
- craniofacial development protein 1
- Gene Aliases
- BCNT, BUCENTAUR, CENP-29, CP27, SWC5, Yeti, p97
- Chromosome Location
- Chr. 16: 75293710 - 75433512 on Build GRCh38
- UniGene ID
- Hs.461361
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006324.2 |
NP_006315.1
|
4, 5 |
675 |
|
XM_011522814.1 |
XP_011521116.1
|
4, 5 |
698 |
|
XM_011522815.1 |
XP_011521117.1
|
4, 5 |
698 |
|
GenBank mRNA |
AB009285.1 |
- |
4, 5 |
647 |
|
AK226031.1 |
- |
5, 6 |
663 |
|
AK313948.1 |
- |
4, 5 |
617 |
|
BC000991.2 |
- |
4, 5 |
671 |
3449836 |
BT009819.1 |
- |
4, 5 |
538 |
|
CR542111.1 |
- |
4, 5 |
538 |
|
D85939.1 |
- |
4, 5 |
647 |
|
D86549.1 |
- |
3, 4 |
474 |
|
LC057679.1 |
- |
4, 5 |
538 |
|
LC057680.1 |
- |
3, 4 |
420 |
|
LC057681.1 |
- |
2, 3 |
292 |
|
LC057682.1 |
- |
4, 5 |
538 |
|
LC057683.1 |
- |
4, 5 |
538 |
|
LC057684.1 |
- |
3, 4 |
420 |
|
View Full Product Details