Gene Transcripts

Gene Symbol
Josd2
Entrez Gene ID
66124
Gene Name
Josephin domain containing 2
Gene Aliases
1110007C05Rik
Chromosome Location
Chr. 7: 44467980 - 44471658 on Build GRCm38
UniGene ID
Mm.358691
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001205070.1 NP_001191999.1 4 534
NM_001205071.1 NP_001192000.1 3 422
NM_001205072.1 NP_001192001.1 3 391
NM_001205073.1 NP_001192002.1 3 388
NM_025368.4 NP_079644.1 3 419
GenBank mRNA AK002872.1 - 3 260
AK003525.1 - 3 344
BC018244.1 - 4 363 4224952
CA326008.1 - 3 333

View Full Product Details