Gene Transcripts

Gene Symbol
Trappc6a
Entrez Gene ID
67091
Gene Name
trafficking protein particle complex 6A
Gene Aliases
1810073E21Rik, 4930519D19Rik, AI480686, TRS33, mhyp
Chromosome Location
Chr. 7: 19507406 - 19516145 on Build GRCm38
UniGene ID
Mm.44213
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025960.3 NP_080236.2 6 735
GenBank mRNA AK007965.1 - 6 734
AK008022.1 - 6 735
AK008462.2 - 6 734
AK088832.1 - 6 734
BC020128.1 - 5 684
BC037154.1 - 6 772 2811950
BC106114.1 - 6 633 4168174

View Full Product Details