Gene Transcripts

Gene Symbol
Rasip1
Entrez Gene ID
69903
Gene Name
Ras interacting protein 1
Gene Aliases
1110025D03Rik, 2610025P08Rik, AI853551, Rain
Chromosome Location
Chr. 7: 45627537 - 45639092 on Build GRCm38
UniGene ID
Mm.181396
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028544.1 NP_082820.1 12 2838
GenBank mRNA AK011557.2 - 6 921
AK033276.1 - 10 2126
AK156602.1 - 12 2850
BC072584.1 - 9 1792
BC127059.1 - 10 1764 40109852
BC150766.1 - 12 2838 9087677

View Full Product Details