Gene Transcripts

Gene Symbol
Mier1
Entrez Gene ID
71148
Gene Name
mesoderm induction early response 1 homolog (Xenopus laevis
Gene Aliases
4933425I22Rik, 5830411K19Rik, er1
Chromosome Location
Chr. 4: 103114390 - 103165754 on Build GRCm38
UniGene ID
Mm.288721
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039081.2 NP_001034170.1 8 803
NM_001286221.1 NP_001273150.1 10 934
NM_001286222.1 NP_001273151.1 10 988
NM_001286223.1 NP_001273152.1 8 784
NM_027696.3 NP_081972.2 10 991
GenBank mRNA AK016915.1 - 2 233
AK129405.1 - 10 911
AY769435.1 - 9 783
AY769436.1 - 8 706
AY769437.1 - 8 756
AY769438.1 - 10 856
BC108273.1 - 8 770 30080243
BC130227.1 - 8 755 40141863
BQ562973.1 - 5 381
CN538186.1 - 5 404
CN701986.1 - 2 157

View Full Product Details