Gene Transcripts

Gene Symbol
Rnd3
Entrez Gene ID
74194
Gene Name
Rho family GTPase 3
Gene Aliases
2610017M01Rik, AI661404, Arhe, Rhoe
Chromosome Location
Chr. 2: 51130439 - 51149111 on Build GRCm38
UniGene ID
Mm.46497
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028810.2 NP_083086.1 5 922
GenBank mRNA AK011442.1 - 5 920
AK035195.1 - 5 907
AK048635.1 - 4 2457
BC009002.1 - 5 892 3500586

View Full Product Details