Gene Transcripts

Gene Symbol
Rabep1
Entrez Gene ID
54189
Gene Name
rabaptin, RAB GTPase binding effector protein 1
Gene Aliases
Rab5ep, rabaptin-5, rabaptin-5alpha
Chromosome Location
Chr. 11: 70844723 - 70943105 on Build GRCm38
UniGene ID
Mm.7087
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291141.1 NP_001278070.1 17 2621
NM_001291142.1 NP_001278071.1 17 2630
NM_001291143.1 NP_001278072.1 19 2821
NM_019400.3 NP_062273.2 18 2750
GenBank mRNA AB001750.1 - 19 2657
AF248489.1 - 17 2416
AF248490.1 - 17 2407
AK134587.1 - 18 2751
BC003921.1 - 8 1756
BC060166.1 - 17 3300
BC080274.1 - 18 2722
BC129853.1 - 18 2575 40109688
BC129854.1 - 18 2569 40109689
D86066.1 - 18 2586

View Full Product Details