Gene Transcripts

Gene Symbol
TRAFD1
Entrez Gene ID
10906
Gene Name
TRAF-type zinc finger domain containing 1
Gene Aliases
FLN29
Chromosome Location
Chr. 12: 112125545 - 112153604 on Build GRCh38
UniGene ID
Hs.5148
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143906.1 NP_001137378.1 10 1978
NM_006700.2 NP_006691.1 10 1447
GenBank mRNA AB007447.1 - 10 1415
AK122620.1 - 10 1962
AK291331.1 - 10 1432
AK312601.1 - 10 1447
BC003553.2 - 9 1373 3547039

View Full Product Details