Gene Transcripts

Gene Symbol
SUB1
Entrez Gene ID
10923
Gene Name
SUB1 homolog, transcriptional regulator
Gene Aliases
P15, PC4, p14
Chromosome Location
Chr. 5: 32531778 - 32604079 on Build GRCh38
UniGene ID
Hs.229641
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006713.3 NP_006704.3 4 346
XM_011513944.2 XP_011512246.1 5 599
XM_017008986.1 XP_016864475.1 5 442
XM_017008987.1 XP_016864476.1 5 319
GenBank mRNA AB209285.1 - 4 241
AK294124.1 - 4 288
AK311754.1 - 4 288
BC009610.1 - 4 274 4297490
BC010537.1 - 4 249 3457167
BC018189.1 - 4 268 3881373
BC022339.1 - 4 279 4134117
BC063402.1 - 5 564
BC071655.1 - 4 263
BC105972.1 - 4 244 3907729
BX472106.1 - 4 347
BX537584.1 - 1 4069
CR456902.1 - 3 218
U12979.1 - 3 218
X79805.1 - 3 218

View Full Product Details