Gene Transcripts
- Gene Symbol
- TMED10
- Entrez Gene ID
- 10972
- Gene Name
- transmembrane p24 trafficking protein 10
- Gene Aliases
- P24(DELTA), S31I125, S31III125, TMP21, Tmp-21-I, p23, p24d1
- Chromosome Location
- Chr. 14: 75131468 - 75176646 on Build GRCh38
- UniGene ID
- Hs.74137
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006827.5 |
NP_006818.3
|
3, 4 |
480 |
|
GenBank mRNA |
AJ004913.1 |
- |
3, 4 |
424 |
|
AK222972.1 |
- |
3, 4 |
432 |
|
AK302920.1 |
- |
2, 3 |
141 |
|
AK308864.1 |
- |
1, 2 |
99 |
|
AK312384.1 |
- |
3, 4 |
449 |
|
AL832012.2 |
- |
3, 4 |
424 |
|
BC001496.2 |
- |
3, 4 |
465 |
2959718 |
BC001825.2 |
- |
3, 4 |
465 |
2959718 |
BX248754.1 |
- |
3, 4 |
431 |
|
BX504393.1 |
- |
3, 4 |
481 |
|
X97442.1 |
- |
3, 4 |
424 |
|
View Full Product Details