Gene Transcripts
- Gene Symbol
- DDX52
- Entrez Gene ID
- 11056
- Gene Name
- DEAD-box helicase 52
- Gene Aliases
- HUSSY19, ROK1
- Chromosome Location
- Chr. 17: 37609739 - 37643464 on Build GRCh38
- UniGene ID
- Hs.590937
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001291476.1 |
NP_001278405.1
|
1, 2 |
128 |
|
NM_007010.4 |
NP_008941.3
|
1, 2 |
128 |
|
XM_011524232.2 |
XP_011522534.1
|
1, 2 |
112 |
|
GenBank mRNA |
AB208938.1 |
- |
1, 2 |
82 |
|
AF077033.1 |
- |
1, 2 |
98 |
|
AK001652.1 |
- |
1, 2 |
109 |
|
AK093661.1 |
- |
1, 2 |
110 |
|
AU132775.1 |
- |
1, 2 |
109 |
|
BC041785.1 |
- |
1, 2 |
110 |
4808288 |
BX492013.1 |
- |
1, 2 |
129 |
|
DA356164.1 |
- |
1, 2 |
128 |
|
View Full Product Details