Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007055.3 | NP_008986.2 | 7, 8 | 1186 | |
GenBank mRNA | AF021351.1 | - | 8, 9 | 1053 | |
AK300329.1 | - | 7, 8 | 1166 | ||
AY091459.1 | - | 7, 8 | 1053 | ||
BC041089.1 | - | 7, 8 | 1147 | 3915501 |