Gene Transcripts
- Gene Symbol
- Trav14-1
- Entrez Gene ID
- 436467
- Gene Name
- T cell receptor alpha variable 14-1
- Gene Aliases
- EG436467, Gm5770
- Chromosome Location
- Chr. 14: 53554057 - 53554558 on Build GRCm38
- UniGene ID
- Mm.359466
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC147823.1 |
- |
2 |
198 |
|
BC147827.1 |
- |
2 |
198 |
|
- Gene Symbol
- Trav14d-2
- Entrez Gene ID
- 100043214
- Gene Name
- T cell receptor alpha variable 14D-2
- Gene Aliases
- EG667553, Gm16454, Tcra-V2.1
- Chromosome Location
- Chr. 14: 52948668 - 52949169 on Build GRCm38
- UniGene ID
- Mm.425908
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
U07881.1 |
- |
2 |
186 |
|
- Gene Symbol
- Trav14n-1
- Entrez Gene ID
- 100043204
- Gene Name
- T cell receptor alpha variable 14N-1
- Gene Aliases
- ENSMUSG00000072532, ENSMUSG00000076804, Gm10896
- Chromosome Location
- Chr. 14: 53166677 - 53167178 on Build GRCm38
- UniGene ID
- Mm.486824
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK039793.1 |
- |
|
215 |
|
- Gene Symbol
- Trav14n-2
- Entrez Gene ID
- 193574
- Gene Name
- T cell receptor alpha variable 14N-2
- Gene Aliases
- EG193574, Gm4743
- Chromosome Location
- Chr. 14: 53259678 - 53260179 on Build GRCm38
- UniGene ID
- Mm.425908
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC147671.1 |
- |
2 |
213 |
|
BC147676.1 |
- |
2 |
213 |
|
BC147840.1 |
- |
2 |
248 |
|
View Full Product Details