Gene Transcripts
- Gene Symbol
- Rps29
- Entrez Gene ID
- 25348
- Gene Name
- ribosomal protein S29
- Gene Aliases
- Chromosome Location
- Chr. 6: 91455333 - 91456709 on Build Rnor_6.0
- UniGene ID
- Rn.34942
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012876.1 |
NP_037008.1
|
2, 3 |
197 |
|
GenBank mRNA |
BC058150.1 |
- |
2, 3 |
195 |
|
FQ209501.1 |
- |
2, 3 |
191 |
|
FQ210507.1 |
- |
2, 3 |
191 |
|
FQ211740.1 |
- |
2, 3 |
191 |
|
FQ216880.1 |
- |
2, 3 |
191 |
|
FQ217140.1 |
- |
2, 3 |
191 |
|
FQ217213.1 |
- |
2, 3 |
191 |
|
FQ218006.1 |
- |
2, 3 |
191 |
|
FQ218037.1 |
- |
2, 3 |
192 |
|
FQ218769.1 |
- |
2, 3 |
191 |
|
FQ221030.1 |
- |
2, 3 |
191 |
|
FQ221251.1 |
- |
2, 3 |
191 |
|
FQ221298.1 |
- |
2, 3 |
191 |
|
FQ221307.1 |
- |
2, 3 |
188 |
|
FQ221360.1 |
- |
2, 3 |
191 |
|
FQ221457.1 |
- |
2, 3 |
191 |
|
FQ221523.1 |
- |
2, 3 |
191 |
|
FQ221587.1 |
- |
2, 3 |
192 |
|
FQ221640.1 |
- |
2, 3 |
191 |
|
FQ221661.1 |
- |
2, 3 |
191 |
|
FQ221707.1 |
- |
2, 3 |
191 |
|
FQ221875.1 |
- |
2, 3 |
191 |
|
FQ221942.1 |
- |
2, 3 |
191 |
|
FQ222143.1 |
- |
2, 3 |
191 |
|
FQ222293.1 |
- |
2, 3 |
191 |
|
FQ222368.1 |
- |
2, 3 |
191 |
|
FQ222566.1 |
- |
2, 3 |
191 |
|
FQ222711.1 |
- |
2, 3 |
191 |
|
FQ222937.1 |
- |
2, 3 |
191 |
|
FQ223071.1 |
- |
2, 3 |
191 |
|
FQ223222.1 |
- |
2, 3 |
191 |
|
FQ223289.1 |
- |
2, 3 |
191 |
|
FQ223378.1 |
- |
2, 3 |
191 |
|
FQ223402.1 |
- |
2, 3 |
191 |
|
FQ223885.1 |
- |
2, 3 |
191 |
|
FQ223975.1 |
- |
2, 3 |
191 |
|
FQ224325.1 |
- |
2, 3 |
191 |
|
FQ224400.1 |
- |
2, 3 |
191 |
|
FQ224465.1 |
- |
2, 3 |
191 |
|
FQ224476.1 |
- |
2, 3 |
191 |
|
FQ224629.1 |
- |
2, 3 |
198 |
|
FQ224661.1 |
- |
2, 3 |
192 |
|
FQ224820.1 |
- |
2, 3 |
191 |
|
FQ228434.1 |
- |
2, 3 |
191 |
|
FQ228730.1 |
- |
2, 3 |
191 |
|
X59051.1 |
- |
2, 3 |
197 |
|
View Full Product Details