Gene Transcripts

Gene Symbol
Scrn2
Entrez Gene ID
360612
Gene Name
secernin 2
Gene Aliases
Chromosome Location
Chr. 10: 84961439 - 84971022 on Build Rnor_6.0
UniGene ID
Rn.15274
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012142.1 NP_001012142.1 3 325
XM_006247342.2 XP_006247404.1 3 554
XM_008768079.1 XP_008766301.1 3 1838
GenBank mRNA BC078939.1 - 3 325

View Full Product Details