Gene Transcripts

Gene Symbol
Sumf2
Entrez Gene ID
360800
Gene Name
sulfatase modifying factor 2
Gene Aliases
Chromosome Location
Chr. 12: 30465025 - 30490683 on Build Rnor_6.0
UniGene ID
Rn.98463
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025125.2 NP_001020296.2 3 313
GenBank mRNA BC091176.1 - 3 319

View Full Product Details