Gene Transcripts

Gene Symbol
Bmp7
Entrez Gene ID
85272
Gene Name
bone morphogenetic protein 7
Gene Aliases
BMP-7
Chromosome Location
Chr. 3: 170879977 - 170955820 on Build Rnor_6.0
UniGene ID
Rn.18030
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191856.2 NP_001178785.1 7 1628
GenBank mRNA AF100787.1 - 7 873
D29769.1 - 4 379

View Full Product Details