Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_005267510.1 | XP_005267567.1 | 5 | 1948 | |
GenBank mRNA | AF131760.1 | - | 1 | 161 | |
AF172816.1 | - | 1 | 833 | ||
AF177336.1 | - | 1 | 275 | ||
AK291030.1 | - | 5 | 1378 | ||
AL834518.1 | - | 5 | 1383 | ||
AY358399.1 | - | 5 | 1778 | ||
BC018816.2 | - | 1 | 343 | ||
BC064901.1 | - | 2 | 937 | ||
BC088378.1 | - | 1 | 226 | ||
BC113714.1 | - | 5 | 1260 | 8322766 | |
BC113716.1 | - | 5 | 1260 | 8322768 | |
BX161500.1 | - | 5 | 1706 |