Gene Transcripts

Gene Symbol
PEX1
Entrez Gene ID
5189
Gene Name
peroxisomal biogenesis factor 1
Gene Aliases
HMLR1, PBD1A, PBD1B, ZWS, ZWS1
Chromosome Location
Chr. 7: 92487023 - 92528531 on Build GRCh38
UniGene ID
Hs.164682
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000466.2 NP_000457.1 6, 7 1450
NM_001282677.1 NP_001269606.1 6, 7 1450
NM_001282678.1 NP_001269607.1 6, 7 1485
XM_017012319.1 XP_016867808.1 2, 3 453
GenBank mRNA AB008112.1 - 6, 7 1421
AB052090.1 - 6, 7 1354
AB052091.1 - 6, 7 1354
AB052092.1 - 6, 7 1354
AB052093.1 - 6, 7 1354
AB052094.1 - 6, 7 1354
AF026086.1 - 6, 7 1414
AF030356.1 - 6, 7 1358
AK292955.1 - 6, 7 1355
AK293757.1 - 6, 7 1393
AK293848.1 - 2, 3 255
AK295686.1 - 3, 4 394
BC035575.1 - 6, 7 1392 5498274

View Full Product Details